CH4102151325 ChemBridge Corporation

3-(3-methylpiperidin-1-yl)propanoic acid

  • MDL number MFCD06753040
  • Cas Number : 858444-38-9
  • Molecular Weight : 171.2368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017724

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4017724
Availability details for
Quantity
  • Availability
  • Add to Cart