CH4102151328 ChemBridge Corporation

3-pyrrolidin-1-ylpropanoic acid

  • MDL number MFCD04114076
  • Cas Number : 76234-38-3
  • Molecular Weight : 143.1836

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017740

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4017740
Availability details for
Quantity
  • Availability
  • Add to Cart