CH4102151329 ChemBridge Corporation

N-(4-oxo-1-adamantyl)acetamide

  • MDL number MFCD01825779
  • Cas Number : 16790-59-3
  • Molecular Weight : 207.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012469

Safety Information

MSDS https://www.hit2lead.com/sds/4012469_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart