CH4102151376 ChemBridge Corporation

2-methyl-2,4,5,6-tetrahydrocyclopenta[c]pyrazol-3-amine

  • MDL number MFCD08444199
  • Cas Number : 885529-68-0
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4019184

Safety Information

MSDS https://www.hit2lead.com/sds/4019184_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart