CH4102151379 ChemBridge Corporation

2-(chloromethyl)-5-isopropyl-1,3,4-oxadiazole

  • MDL number MFCD08059899
  • Cas Number : 3914-46-3
  • Molecular Weight : 160.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(chloromethyl)-5-isopropyl-1,3,4-oxadiazole
Inchi Code 1S/C6H9ClN2O/c1-4(2)6-9-8-5(3-7)10-6/h4H,3H2,1-2H3
Inchi Key UCGJAQHFSXERIT-UHFFFAOYSA-N

Description

Purity 95%
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4012860

Safety Information

MSDS https://www.hit2lead.com/sds/4012860_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart