CH4102151406 ChemBridge Corporation

3-(2-amino-2-oxoethoxy)benzoic acid

  • MDL number MFCD04580891
  • Cas Number : 869942-82-5
  • Molecular Weight : 195.1721

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4021971

Safety Information

MSDS https://www.hit2lead.com/sds/4021971_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart