CH4102151447 ChemBridge Corporation

6-bromoquinazolin-4-amine

  • MDL number MFCD02228849
  • Cas Number : 21419-48-7
  • Molecular Weight : 224.0573

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4022407

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4022407
Availability details for
Quantity
  • Availability
  • Add to Cart