CH4102151478 ChemBridge Corporation

3-pyrrolidin-1-ylpropan-1-ol

  • MDL number MFCD01687769
  • Cas Number : 19748-66-4
  • Molecular Weight : 129.2001

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012899

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4012899
Availability details for
Quantity
  • Availability
  • Add to Cart