CH4102151491 ChemBridge Corporation

2-methyl-1-prop-2-yn-1-yl-1H-indole-3-carbaldehyde

  • MDL number MFCD04342457
  • Cas Number : 842973-82-4
  • Molecular Weight : 197.2325

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4022978

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4022978
Availability details for
Quantity
  • Availability
  • Add to Cart