CH4102151537 ChemBridge Corporation

4-(allyloxy)aniline

  • MDL number MFCD01902260
  • Cas Number : 1688-69-3
  • Molecular Weight : 149.1897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023587

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4023587
Availability details for
Quantity
  • Availability
  • Add to Cart