CH4102151664 ChemBridge Corporation

2-(2-sec-butylphenoxy)butanoic acid

  • MDL number MFCD08752996
  • Cas Number : 915921-89-0
  • Molecular Weight : 236.3068

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024055

Safety Information

MSDS https://www.hit2lead.com/sds/4024055_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart