CH4102151705 ChemBridge Corporation

(4-isothiocyanatophenyl)(2-methoxyphenyl)amine

  • MDL number MFCD08060078
  • Cas Number : 915919-57-2
  • Molecular Weight : 256.3229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4150306

Safety Information

MSDS https://www.hit2lead.com/sds/4150306_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart