CH4102151722 ChemBridge Corporation

1-(3-bromopropoxy)-3-methoxybenzene

  • MDL number MFCD00596670
  • Cas Number : 3245-40-7
  • Molecular Weight : 245.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5300128

Safety Information

MSDS https://www.hit2lead.com/sds/5300128_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart