CH4102151723 ChemBridge Corporation

4-{[6-(1H-pyrazol-1-yl)-4-pyrimidinyl]oxy}benzaldehyde

  • MDL number MFCD00299070
  • Cas Number : 1015846-03-3
  • Molecular Weight : 266.2548

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5339663

Safety Information

MSDS https://www.hit2lead.com/sds/5339663_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart