CH4102151726 ChemBridge Corporation

4-(3-chloropropanoyl)-2,6-dimethylmorpholine

  • MDL number MFCD01213673
  • Cas Number : 915920-51-3
  • Molecular Weight : 205.6818

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5558519

Safety Information

MSDS https://www.hit2lead.com/sds/5558519_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart