CH4102151728 ChemBridge Corporation

1-(2-bromoethoxy)-2-isopropylbenzene

  • MDL number MFCD02030521
  • Cas Number : 3245-44-1
  • Molecular Weight : 243.1402

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5581725

Safety Information

MSDS https://www.hit2lead.com/sds/5581725_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart