CH4102151734 ChemBridge Corporation

3-(1H-1,2,4-triazol-1-yl)-1-adamantanamine

  • MDL number MFCD08691741
  • Cas Number : 915920-86-4
  • Molecular Weight : 218.2981

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5636113

Safety Information

MSDS https://www.hit2lead.com/sds/5636113_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart