CH4102151754 ChemBridge Corporation

8-hydroxy-2,3-dihydrocyclopenta[c]chromen-4(1H)-one

  • MDL number MFCD08691761
  • Cas Number : 131526-88-0
  • Molecular Weight : 202.206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6139319

Safety Information

MSDS https://www.hit2lead.com/sds/6139319_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart