CH4102151766 ChemBridge Corporation

4-(bromoacetyl)morpholine

  • MDL number MFCD06800306
  • Cas Number : 40299-87-4
  • Molecular Weight : 208.0531

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023671

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4023671
Availability details for
Quantity
  • Availability
  • Add to Cart