CH4102151768 ChemBridge Corporation

isopropyl 3-aminobenzoate

  • MDL number MFCD07601158
  • Cas Number : 35005-25-5
  • Molecular Weight : 179.2157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024620

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4024620
Availability details for
Quantity
  • Availability
  • Add to Cart