CH4102151798 ChemBridge Corporation

3-[(4-methylphenyl)amino]-3-oxopropanoic acid

  • MDL number MFCD01794761
  • Cas Number : 95262-01-4
  • Molecular Weight : 193.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024704

Safety Information

MSDS https://www.hit2lead.com/sds/4024704_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart