CH4102151862 ChemBridge Corporation

N-methyl-1-[1-(2-methylphenyl)-1H-tetrazol-5-yl]-1-phenylmethanamine

  • MDL number MFCD01821143
  • Cas Number : 400747-21-9
  • Molecular Weight : 279.3397

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100003

Safety Information

MSDS https://www.hit2lead.com/sds/4100003_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart