CH4102151897 ChemBridge Corporation

N-methyl-1-(2-thienyl)methanamine

  • MDL number MFCD04625435
  • Cas Number : 58255-18-8
  • Molecular Weight : 127.2074

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100657

Safety Information

MSDS https://www.hit2lead.com/sds/4100657_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart