CH4102151904 ChemBridge Corporation

1,3-dihydro-2-benzofuran-5-amine

  • MDL number MFCD05864561
  • Cas Number : 61964-08-7
  • Molecular Weight : 135.1632

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4100713

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4100713
Availability details for
Quantity
  • Availability
  • Add to Cart