CH4102151931 ChemBridge Corporation

1-(1-methyl-1,2,3,4-tetrahydroquinolin-6-yl)methanamine

  • MDL number MFCD05864543
  • Cas Number : 797807-54-6
  • Molecular Weight : 176.2581

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101141

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4101141
Availability details for
Quantity
  • Availability
  • Add to Cart