CH4102151938 ChemBridge Corporation

1-(1-methyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD02090924
  • Cas Number : 400877-05-6
  • Molecular Weight : 111.1451

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101278

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4101278
Availability details for
Quantity
  • Availability
  • Add to Cart