CH4102151962 ChemBridge Corporation

6-methoxy-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD01717129
  • Cas Number : 42923-77-3
  • Molecular Weight : 163.2163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4101917

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4101917
Availability details for
Quantity
  • Availability
  • Add to Cart