CH4102151998 ChemBridge Corporation

4-(4-cyclohexylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD01993687
  • Cas Number : 105512-86-5
  • Molecular Weight : 258.3818

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4102892

Safety Information

MSDS https://www.hit2lead.com/sds/4102892_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart