CH4102152004 ChemBridge Corporation

N-methyl-1-(1-methylpiperidin-4-yl)methanamine

  • MDL number MFCD06738983
  • Cas Number : 405928-19-0
  • Molecular Weight : 142.2419

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4103004

Safety Information

MSDS https://www.hit2lead.com/sds/4103004_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart