CH4102152005 ChemBridge Corporation

N-methyl-1-(1-methylpiperidin-3-yl)methanamine

  • MDL number MFCD05864531
  • Cas Number : 639078-61-8
  • Molecular Weight : 142.2419

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4103005

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4103005
Availability details for
Quantity
  • Availability
  • Add to Cart