CH4102152025 ChemBridge Corporation

2-amino-4-ethyl-5-methylthiophene-3-carboxamide

  • MDL number MFCD01114969
  • Cas Number : 350996-89-3
  • Molecular Weight : 184.2587

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4133061

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4133061
Availability details for
Quantity
  • Availability
  • Add to Cart