CH4102152036 ChemBridge Corporation

2-methyl-N-phenylalanine

  • MDL number MFCD05228616
  • Cas Number : 59081-61-7
  • Molecular Weight : 179.2157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4140261

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4140261_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart