CH4102152052 ChemBridge Corporation

2-bromo-N-phenylacetamide

  • MDL number MFCD00462518
  • Cas Number : 5326-87-4
  • Molecular Weight : 214.0592

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023663

Safety Information

MSDS https://www.hit2lead.com/sds/4023663_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart