CH4102152053 ChemBridge Corporation

N-(1,1-dioxido-2,3-dihydro-3-thienyl)glycine

  • MDL number MFCD00195978
  • Cas Number : 201990-24-1
  • Molecular Weight : 191.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4140771

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4140771
Availability details for
Quantity
  • Availability
  • Add to Cart