CH4102152062 ChemBridge Corporation

2-(2-sec-butylphenoxy)propanoic acid

  • MDL number MFCD08559345
  • Cas Number : 887029-70-1
  • Molecular Weight : 222.2802

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024052

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4024052
Availability details for
Quantity
  • Availability
  • Add to Cart