CH4102152084 ChemBridge Corporation

{2-[(4-oxopiperidin-1-yl)methyl]phenyl}boronic acid

  • MDL number MFCD05864584
  • Cas Number : 697739-42-7
  • Molecular Weight : 233.0713

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4201136

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4201136_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart