CH4102152093 ChemBridge Corporation

2-bromo-N-(2,4-dimethylphenyl)acetamide

  • MDL number MFCD06149118
  • Cas Number : 255909-04-7
  • Molecular Weight : 242.1124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023665

Safety Information

MSDS https://www.hit2lead.com/sds/4023665_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart