CH4102152104 ChemBridge Corporation

3-(4-methoxyphenyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD01133696
  • Cas Number : 199682-73-0
  • Molecular Weight : 202.2093

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4300297

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4300297
Availability details for
Quantity
  • Availability
  • Add to Cart