CH4102152115 ChemBridge Corporation

4-phenyl-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD02665589
  • Cas Number : 75390-44-2
  • Molecular Weight : 189.2337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4301057

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4301057
Availability details for
Quantity
  • Availability
  • Add to Cart