CH4102152140 ChemBridge Corporation

5-(4H-1,2,4-triazol-3-ylthio)-2-furaldehyde

  • MDL number MFCD18839918
  • Cas Number : 797807-53-5
  • Molecular Weight : 195.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4302952

Safety Information

MSDS https://www.hit2lead.com/sds/4302952_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart