CH4102152161 ChemBridge Corporation

5-methoxy-1,2-dimethyl-1H-indole-3-carboxylic acid

  • MDL number MFCD00458840
  • Cas Number : 105909-93-1
  • Molecular Weight : 219.2365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400196

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4400196
Availability details for
Quantity
  • Availability
  • Add to Cart