CH4102152162 ChemBridge Corporation

2-(4-methylphenyl)quinoline-4-carboxylic acid

  • MDL number MFCD00047598
  • Cas Number : 20389-05-3
  • Molecular Weight : 263.2906

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400199

Safety Information

MSDS https://www.hit2lead.com/sds/4400199_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart