CH4102152184 ChemBridge Corporation

6-(4-methoxyphenyl)-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD02926405
  • Cas Number : 147269-07-6
  • Molecular Weight : 245.2307

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400284

Safety Information

MSDS https://www.hit2lead.com/sds/4400284_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart