CH4102152188 ChemBridge Corporation

2-[(4,6-dimethylpyrimidin-2-yl)thio]propanoic acid

  • MDL number MFCD02244111
  • Cas Number : 433242-31-0
  • Molecular Weight : 212.2688

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400367

Safety Information

MSDS https://www.hit2lead.com/sds/4400367_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart