CH4102152210 ChemBridge Corporation

9-methyl-9H-carbazole-3-carboxylic acid

  • MDL number MFCD00813989
  • Cas Number : 89374-79-8
  • Molecular Weight : 225.2426

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401031

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4401031
Availability details for
Quantity
  • Availability
  • Add to Cart