CH4102152231 ChemBridge Corporation

1,2,5-oxadiazole-3-carboxylic acid

  • MDL number MFCD00604386
  • Cas Number : 88598-08-7
  • Molecular Weight : 114.0596

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401609

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4401609
Availability details for
Quantity
  • Availability
  • Add to Cart