CH4102152236 ChemBridge Corporation

1-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD01463245
  • Cas Number : 18471-99-3
  • Molecular Weight : 203.1941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401867

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4401867
Availability details for
Quantity
  • Availability
  • Add to Cart