CH4102152239 ChemBridge Corporation

5-(4-methoxyphenyl)-2-furoic acid

  • MDL number MFCD03937477
  • Cas Number : 52938-99-5
  • Molecular Weight : 218.2054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401950

Safety Information

MSDS https://www.hit2lead.com/sds/4401950_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart