CH4102152240 ChemBridge Corporation

5-(methoxymethyl)-2-furoic acid

  • MDL number MFCD04367609
  • Cas Number : 1917-60-8
  • Molecular Weight : 156.136

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401976

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4401976
Availability details for
Quantity
  • Availability
  • Add to Cart