CH4102152256 ChemBridge Corporation

1-methyl-3-propyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD02169703
  • Cas Number : 139755-99-0
  • Molecular Weight : 168.1931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4402460

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4402460
Availability details for
Quantity
  • Availability
  • Add to Cart